2-Fluoro-6-methoxypyridin-4-amine 95%

Supplier: Ambeed
Warning

A524423-250MG A524423-50MG
77144-204EA 453.01 USD
77144-204 77144-206
2-Fluoro-6-methoxypyridin-4-amine 95%
2-Fluoro-6-methoxypyridin-4-amine

Precaución: for Research Use Only

Formula: C₆H₇FN₂O
MW: 142.13 g/mol
MDL Number: MFCD16606163
CAS Number: 1227586-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR