(S)-1-Methyl-3-(hydroxymethyl)piperidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-(1-Methylpiperidin-3-yl)methanol, [(S)-1-Methylpiperidin-3-yl]methanol
A523660-250MG
A523660-5G
77144-194EA
23.68
USD
77144-194
77144-196
(S)-1-Methyl-3-(hydroxymethyl)piperidine 98%
(S)-1-Methyl-3-(hydroxymethyl)piperidine
Precaución: for Research Use Only
Formula:
C₇H₁₅NO |
MDL Number:
MFCD11035736 CAS Number: 205194-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...