2,3,5,6,7,8-Hexahydro-1H-cyclopenta[b]quinolin-9-amine hydrochloride hydrate 97%

Supplier: Ambeed
Warning

A523625-250MG A523625-5G
77144-190EA 13.39 USD
77144-190 77144-192
2,3,5,6,7,8-Hexahydro-1H-cyclopenta[b]quinolin-9-amine hydrochloride hydrate 97%
2,3,5,6,7,8-Hexahydro-1H-cyclopenta[b]quinolin-9-amine hydrochloride hydrate

Precaución: for Research Use Only

Formula: C₁₂H₁₉ClN₂O
MDL Number: MFCD00517543
CAS Number: 118499-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR