(S)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (S)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene

A522831-1G A522831-250MG
77144-178EA 227.04 USD
77144-178 77144-180
(S)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl ≥98%, ee 99%
(S)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl

Precaución: for Research Use Only

Formula: C₂₂H₁₆Br₂O₂
MW: 472.18 g/mol
MDL Number: MFCD04038415
CAS Number: 75714-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR