6-Bromo-1-isopropyl-1H-pyrrolo[3,2-c]pyridine 97%

Supplier: Ambeed
Danger

A522630-1G A522630-250MG
77144-174EA 242.95 USD
77144-174 77144-176
6-Bromo-1-isopropyl-1H-pyrrolo[3,2-c]pyridine 97%
6-Bromo-1-isopropyl-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrN₂
MW: 239.11 g/mol
MDL Number: MFCD29921884
CAS Number: 1612172-57-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR