Potassium trifluoro(4-methylthiophen-2-yl)borate 95%

Supplier: Ambeed
Warning

A522542-1G A522542-250MG A522542-5G
77144-168EA 53.05 USD
77144-168 77144-170 77144-172
Potassium trifluoro(4-methylthiophen-2-yl)borate 95%
Potassium trifluoro(4-methylthiophen-2-yl)borate

Precaución: for Research Use Only

Formula: C₅H₅BF₃KS
MW: 204.06 g/mol
MDL Number: MFCD11052667
CAS Number: 1111732-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR