(5-Chloro-2-methyl-1H-indol-3-yl)acetic acid 98%

Supplier: Ambeed
Warning

A520108-100MG A520108-1G A520108-250MG
77144-162EA 9.26 USD
77144-162 77144-164 77144-166
(5-Chloro-2-methyl-1H-indol-3-yl)acetic acid 98%
(5-Chloro-2-methyl-1H-indol-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClNO₂
MDL Number: MFCD00152035
CAS Number: 19017-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR