(2-Benzothiazolylthio)acetic acid 95%

Supplier: Ambeed
Danger

Synonyms: [(1,3-Benzothiazol-2-yl)sulphanyl]acetic acid, 2-(Benzo[d]thiazol-2-ylthio)acetic acid

A520033-25G A520033-5G
77144-158EA 24.41 USD
77144-158 77144-160
(2-Benzothiazolylthio)acetic acid 95%
(2-Benzothiazolylthio)acetic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₂S₂
MW: 225.28 g/mol
MDL Number: MFCD00227391
CAS Number: 6295-57-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR