2-(4-Bromo-2-hydroxyphenyl)acetic acid 97%

Supplier: Ambeed
Warning

A518858-100MG A518858-250MG
77144-150EA 13.39 USD
77144-150 77144-152
2-(4-Bromo-2-hydroxyphenyl)acetic acid 97%
2-(4-Bromo-2-hydroxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MDL Number: MFCD18391507
CAS Number: 1261497-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR