2-(3-Bromo-2-methylphenyl)acetonitrile 95%
Supplier: Ambeed
|
Warning
|
A516991-1G
A516991-250MG
A516991-5G
77144-132EA
92.71
USD
77144-132
77144-134
77144-136
2-(3-Bromo-2-methylphenyl)acetonitrile 95%
2-(3-Bromo-2-methylphenyl)acetonitrile
Precaución: for Research Use Only
Formula:
C₉H₈BrN |
MDL Number:
MFCD18392229 CAS Number: 76574-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...