3-Chlorothiobenzamide 98%

Supplier: Ambeed
Warning

A516790-25G A516790-5G
77144-124EA 60.77 USD
77144-124 77144-126
3-Chlorothiobenzamide 98%
3-Chlorothiobenzamide

Precaución: for Research Use Only

Formula: C₇H₆ClNS
MW: 171.65 g/mol
MDL Number: MFCD00276993
CAS Number: 2548-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR