6-Fluoro-3-methoxypyridin-2-amine 97%

Supplier: Ambeed
Warning

A512458-250MG A512458-100MG
77144-110EA 37.07 USD
77144-110 77747-050
6-Fluoro-3-methoxypyridin-2-amine 97%
6-Fluoro-3-methoxypyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₇FN₂O
MDL Number: MFCD28722398
CAS Number: 1805602-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR