Bis(1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene)copper(I) tetrafluoroborate 98%

Supplier: Ambeed
Warning

A510744-100MG A510744-1G A510744-250MG A510744-5G
77144-100EA 31.83 USD
77144-100 77144-102 77144-104 77144-106
Bis(1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene)copper(I) tetrafluoroborate 98%
Bis(1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene)copper(I) tetrafluoroborate

Precaución: for Research Use Only

Formula: C₅₄H₇₂BCuF₄N₄
MW: 927.53 g/mol
MDL Number: MFCD11045422
CAS Number: 886061-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR