(S)-1-Benzyl 4-tert-butyl 2-(hydroxymethyl)piperazine-1,4-dicarboxylate 97%

Supplier: Ambeed
Warning

A510118-100MG A510118-1G A510118-5G
77144-096EA 54.11 USD
77144-096 77144-098 77686-328
(S)-1-Benzyl 4-tert-butyl 2-(hydroxymethyl)piperazine-1,4-dicarboxylate 97%
(S)-1-Benzyl 4-tert-butyl 2-(hydroxymethyl)piperazine-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₆N₂O₅
MW: 350.40 g/mol
MDL Number: MFCD09971767
CAS Number: 529516-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR