1,3-Difluoro-2-iodo-4-nitrobenzene 95%

Supplier: Ambeed
Warning

A509718-1G A509718-250MG
77144-092EA 49.44 USD
77144-092 77144-094
1,3-Difluoro-2-iodo-4-nitrobenzene 95%
1,3-Difluoro-2-iodo-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂F₂INO₂
MDL Number: MFCD28789221
CAS Number: 1145881-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR