2-Bromo-3-chloro-1,4-difluorobenzene 97%

Supplier: Ambeed
Warning

A506634-100MG A506634-250MG A506634-1G A506634-5G
77144-064EA 6.18 USD
77144-064 77144-066 77617-164 77747-044
2-Bromo-3-chloro-1,4-difluorobenzene 97%
2-Bromo-3-chloro-1,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClF₂
MW: 227.43 g/mol
MDL Number: MFCD12922607
CAS Number: 1208077-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR