2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 97%

Supplier: Ambeed
Warning

A505090-1G A505090-5G
77144-060EA 48.41 USD
77144-060 77144-062
2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 97%
2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₇BN₂O₂
MDL Number: MFCD16996233
CAS Number: 934426-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR