1-Cyclopropyl-1H-pyrazol-3-amine 98%

Supplier: Ambeed
Danger

A504372-100MG A504372-1G A504372-250MG A504372-50MG
77144-046EA 43.26 USD
77144-046 77144-048 77144-050 77144-052
1-Cyclopropyl-1H-pyrazol-3-amine 98%
1-Cyclopropyl-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MDL Number: MFCD16810809
CAS Number: 1240565-06-3
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR