Tenovin-3 98%

Supplier: Ambeed
Warning

Synonyms: N-((4-Aminophenyl)carbamothioyl)-4-(tert-butyl)benzamide, N-[(4-aminophenyl)carbamothioyl]-4-tert-butylbenzamide

A501104-100MG A501104-10MG A501104-1MG A501104-25MG A501104-50MG A501104-5MG
77144-028EA 41.38 USD
77144-028 77144-030 77144-032 77144-034 77144-036 77144-038
Tenovin-3 98%
Tenovin-3

Precaución: for Research Use Only

Formula: C₁₈H₂₁N₃OS
MW: 327.44 g/mol
MDL Number: MFCD25976437
CAS Number: 1011301-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR