2-([1,1'-Biphenyl]-2-yloxy)acetic acid 97%

Supplier: Ambeed
Warning

A500788-1G A500788-5G
77144-024EA 41.2 USD
77144-024 77144-026
2-([1,1'-Biphenyl]-2-yloxy)acetic acid 97%
2-([1,1'-Biphenyl]-2-yloxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
MDL Number: MFCD00152752
CAS Number: 5348-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR