2,3-Dichloropyrido[3,4-b]pyrazine 95%

Supplier: Ambeed
Warning

A500474-100MG A500474-1G A500474-250MG
77144-012EA 64.88 USD
77144-012 77144-014 77144-016
2,3-Dichloropyrido[3,4-b]pyrazine 95%
2,3-Dichloropyrido[3,4-b]pyrazine

Precaución: for Research Use Only

Formula: C₇H₃Cl₂N₃
MDL Number: MFCD19374272
CAS Number: 35251-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR