(2-Isobutoxy-4-(trifluoromethyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A499450-1G A499450-250MG A499450-5G
77144-002EA 94.43 USD
77144-002 77144-004 77144-006
(2-Isobutoxy-4-(trifluoromethyl)phenyl)boronic acid 97%
(2-Isobutoxy-4-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄BF₃O₃
MW: 262.03 g/mol
MDL Number: MFCD21609513
CAS Number: 2096331-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR