5-(Cyclopropylaminomethyl)-2-fluorophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

A497291-1G A497291-250MG
77143-996EA 37.14 USD
77143-996 77143-998 77144-000
5-(Cyclopropylaminomethyl)-2-fluorophenylboronic acid pinacol ester 97%
5-(Cyclopropylaminomethyl)-2-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₃BFNO₂
MDL Number: MFCD18434459
CAS Number: 2096338-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR