(2-Mercaptoethyl)trimethylsilane 95%

Supplier: Ambeed
Danger

A496395-1G A496395-5G A496395-250MG
77143-992EA 254.41 USD
77143-992 77143-994 77686-300
(2-Mercaptoethyl)trimethylsilane 95%
(2-Mercaptoethyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₅H₁₄SSi
MDL Number: MFCD00077888
CAS Number: 18143-30-1
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR