N-Boc-4-fluoroaniline 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (4-fluorophenyl)carbamate, 4-Fluoroaniline, N-BOC protected

A494381-100G A494381-1G A494381-25G
77143-978EA 309.79 USD
77143-978 77143-980 77143-982
N-Boc-4-fluoroaniline 98%
N-Boc-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₁₁H₁₄FNO₂
MW: 211.2328 g/mol
MDL Number: MFCD07127707
CAS Number: 60144-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR