2-((6-Phenyl-2,3,4,9-tetrahydro-1H-carbazol-1-yl)amino)ethan-1-ol 98%

Supplier: Ambeed
Warning

Synonyms: CASIN

A490851-10MG A490851-25MG A490851-5MG A490851-1MG
77143-938EA 36.08 USD
77143-938 77143-940 77143-942 77617-114
2-((6-Phenyl-2,3,4,9-tetrahydro-1H-carbazol-1-yl)amino)ethan-1-ol 98%
2-((6-Phenyl-2,3,4,9-tetrahydro-1H-carbazol-1-yl)amino)ethan-1-ol

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₂O
MW: 306.40 g/mol
MDL Number: MFCD01853271
CAS Number: 425399-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR