4-Bromo-2-fluorobenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-fluorobenzyl Alcohol, (4-Bromo-2-fluorophenyl)methanol

A490690-100G A490690-25G A490690-5G
77143-932EA 102.91 USD
77143-932 77143-934 77143-936
4-Bromo-2-fluorobenzyl alcohol 97%
4-Bromo-2-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MW: 205.03 g/mol
MDL Number: MFCD00143265
CAS Number: 188582-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR