1-Ethyl-2,3-dimethylimidazoliumhexafluorophosphate 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Ethyl-2,3-dimethyl-1H-imidazol-3-ium hexafluorophosphate(V)
A490164-100G
A490164-10G
A490164-1G
A490164-25G
A490164-5G
77143-922EA
95.49
USD
77143-922
77143-924
77143-926
77143-928
77143-930
1-Ethyl-2,3-dimethylimidazoliumhexafluorophosphate 98+%
1-Ethyl-2,3-dimethylimidazoliumhexafluorophosphate
Precaución: for Research Use Only
Formula:
C₇H₁₃F₆N₂P MW: 270.16 g/mol |
MDL Number:
MFCD08458503 CAS Number: 292140-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...