5-Isopropyl-1-methyl-1H-imidazole 97%

Supplier: Ambeed
Warning

A489785-1G A489785-5G
77143-918EA 135.8 USD
77143-918 77143-920
5-Isopropyl-1-methyl-1H-imidazole 97%
5-Isopropyl-1-methyl-1H-imidazole

Precaución: for Research Use Only

Formula: C₇H₁₂N₂
MW: 124.18 g/mol
MDL Number: MFCD18829907
CAS Number: 196304-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR