(R)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (R)-(S)-PPFA

A488771-100MG A488771-1G A488771-250MG A488771-5G
77143-910EA 28.65 USD
77143-910 77143-912 77143-914 77143-916
(R)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine ≥98%, ee 99%
(R)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine

Precaución: for Research Use Only

Formula: C₂₆H₂₈FeNP
MW: 441.34 g/mol
MDL Number: MFCD00075286
CAS Number: 74311-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR