4,5-Dimethoxypyrimidine 97%
Supplier: Ambeed
|
Warning
|
A487995-100MG
A487995-250MG
77143-906EA
23.34
USD
77143-906
77143-908
4,5-Dimethoxypyrimidine 97%
4,5-Dimethoxypyrimidine
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₂ MW: 140.14 g/mol |
MDL Number:
MFCD28162964 CAS Number: 71133-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...