(+)-N,N'-(1S,2S)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide) ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: N,N'-((1S,2S)-Cyclohexane-1,2-diyl)dipicolinamide, (S,S)-DACH-Pyridyl Trost Ligand

A487907-100MG A487907-1G A487907-250MG A487907-5G
77143-898EA 13.8 USD
77143-898 77143-900 77143-902 77143-904
(+)-N,N'-(1S,2S)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide) ≥98%, ee 99%
(+)-N,N'-(1S,2S)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide)

Precaución: for Research Use Only

Formula: C₁₈H₂₀N₄O₂
MW: 324.3800 g/mol
MDL Number: MFCD02684544
CAS Number: 172138-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR