1-Butyl-1-methylpyrrolidiniumtrifluormethansulphonate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Butyl-1-methylpyrrolidin-1-ium trifluoromethanesulphonate, 1-Butyl-1-methylpyrrolidinium triflate, 1-Butyl-1-methylpyrrolidinium trifluoromethanesulphonate

A487212-100G A487212-1G A487212-25G A487212-5G
77143-884EA 338.87 USD
77143-884 77143-886 77143-888 77143-890
1-Butyl-1-methylpyrrolidiniumtrifluormethansulphonate 98%
1-Butyl-1-methylpyrrolidiniumtrifluormethansulphonate

Precaución: for Research Use Only

Formula: C₁₀H₂₀F₃NO₃S
MW: 291.33 g/mol
MDL Number: MFCD08458925
CAS Number: 367522-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR