tert-Butyl 5-chloroisoindoline-2-carboxylate 97%

Supplier: Ambeed
Warning

A485767-100MG A485767-1G A485767-250MG
77143-872EA 40.32 USD
77143-872 77143-874 77143-876
tert-Butyl 5-chloroisoindoline-2-carboxylate 97%
tert-Butyl 5-chloroisoindoline-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₆ClNO₂
MW: 253.72 g/mol
MDL Number: MFCD16620845
CAS Number: 871723-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR