2-Bromo-4-iodobenzamide 97%

Supplier: Ambeed
Warning

A485609-100MG A485609-1G A485609-250MG
77143-866EA 45.62 USD
77143-866 77143-868 77143-870
2-Bromo-4-iodobenzamide 97%
2-Bromo-4-iodobenzamide

Precaución: for Research Use Only

Formula: C₇H₅BrINO
MW: 325.92 g/mol
MDL Number: MFCD18391622
CAS Number: 1261516-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR