PEG3-carboxylic acid tert-Butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(2-(2-hydroxyethoxy)ethoxy)propanoate, 3-[2-(2-Hydroxyethoxy)ethoxy]propanoic acid tert-Butyl ester, Hydroxy-PEG2-(CH2)2-Boc, tert-Butyl 3-[2-(2-Hydroxyethoxy)ethoxy]propanoate, 9-Hydroxy-4,7-dioxanonanoic acid tert-Butyl ester, tert-Butyl 9-Hydroxy-4,7-dioxanonanoate

A485329-10G A485329-1G A485329-250MG A485329-25G A485329-5G A485329-100G
77143-856EA 263.11 USD
77143-856 77143-858 77143-860 77143-862 77143-864 77617-100
PEG3-carboxylic acid tert-Butyl ester 95%
PEG3-carboxylic acid tert-Butyl ester

Precaución: for Research Use Only

Formula: C₁₁H₂₂O₅
MW: 234.29 g/mol
MDL Number: MFCD20926380
CAS Number: 133803-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR