2-Carboxy-4-fluorobenzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-Carboxy-4-fluorophenylboronic acid pinacol ester, 5-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

A485326-1G A485326-250MG A485326-5G
77143-850EA 169.95 USD
77143-850 77143-852 77143-854
2-Carboxy-4-fluorobenzeneboronic acid pinacol ester 97%
2-Carboxy-4-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₄
MDL Number: MFCD22414477
CAS Number: 2377607-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR