2-[4-(Trifluoromethoxy)phenyl]-1,3,8-triazaspiro[4.5]dec-1-en-4-one 95%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(Trifluoromethoxy)phenyl)-1,3,8-triazaspiro[4.5]dec-1-en-4-one, 4-(4-Oxo-1,3,8-triazaspiro[4.5]dec-1-en-2-yl)-α,α,α-trifluoroanisole, 4-Oxo-2-[4-(trifluoromethoxy)phenyl]-1,3,8-triazaspiro[4.5]dec-1-ene

A484002-1G A484002-250MG A484002-5G A484002-25G
77143-844EA 56.23 USD
77143-844 77143-846 77143-848 77572-534
2-[4-(Trifluoromethoxy)phenyl]-1,3,8-triazaspiro[4.5]dec-1-en-4-one 95%
2-[4-(Trifluoromethoxy)phenyl]-1,3,8-triazaspiro[4.5]dec-1-en-4-one

Precaución: for Research Use Only

Formula: C₁₄H₁₄F₃N₃O₂
MW: 313.275 g/mol
MDL Number: MFCD22375530
CAS Number: 1253924-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR