4-(Difluoromethoxy)-2-methylthiazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A483053-100MG A483053-250MG
77143-838EA 89.12 USD
77143-838 77143-840
4-(Difluoromethoxy)-2-methylthiazole-5-carboxylic acid 97%
4-(Difluoromethoxy)-2-methylthiazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅F₂NO₃S
MW: 209.17 g/mol
MDL Number: MFCD24483817
CAS Number: 851755-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR