1-Bromo-3,5-dimethoxy-4-fluorobenzene 95%

Supplier: Ambeed
Warning

A482005-10G A482005-1G A482005-250MG A482005-25G A482005-5G
77143-828EA 215.26 USD
77143-828 77143-830 77143-832 77143-834 77143-836
1-Bromo-3,5-dimethoxy-4-fluorobenzene 95%
1-Bromo-3,5-dimethoxy-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂
MDL Number: MFCD28738436
CAS Number: 204654-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR