2-Fluorophenethyl bromide 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Bromoethyl)-2-fluorobenzene

A481846-1G A481846-250MG A481846-5G
77143-822EA 35.02 USD
77143-822 77143-824 77143-826
2-Fluorophenethyl bromide 95%
2-Fluorophenethyl bromide

Precaución: for Research Use Only

Formula: C₈H₈BrF
MW: 203.05 g/mol
MDL Number: MFCD04117466
CAS Number: 91319-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR