2,6-Difluoro-3-iodobenzonitrile 97%

Supplier: Ambeed
Warning

A481645-100MG A481645-1G A481645-250MG
77143-804EA 152.77 USD
77143-804 77143-806 77143-808
2,6-Difluoro-3-iodobenzonitrile 97%
2,6-Difluoro-3-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂F₂IN
MW: 264.99 g/mol
MDL Number: MFCD25542085
CAS Number: 1447606-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR