Methyl 2-bromo-4-chloronicotinate 97%

Supplier: Ambeed
Warning

A479293-100MG A479293-1G A479293-250MG
77143-776EA 77.45 USD
77143-776 77143-778 77143-780
Methyl 2-bromo-4-chloronicotinate 97%
Methyl 2-bromo-4-chloronicotinate

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MW: 250.47 g/mol
MDL Number: MFCD28734551
CAS Number: 1804384-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR