Dicarbonyl(2,4-pentanedionato)rhodium(I) 98% 39% Rh

Supplier: Ambeed
Danger

Synonyms: (Acetylacetonato)dicarbonylrhodium(I), Dicarbonylacetylacetonato rhodium(I)

A479174-250MG A479174-100MG
77143-774EA 92.71 USD
77143-774 77746-986
Dicarbonyl(2,4-pentanedionato)rhodium(I) 98% 39% Rh
Dicarbonyl(2,4-pentanedionato)rhodium(I)

Precaución: for Research Use Only

Formula: C₇H₈O₄Rh
MW: 259.04 g/mol
MDL Number: MFCD00009884
CAS Number: 14874-82-9
UN: 3467
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR