2,6-Dichloro-4-nitroanisole 97%

Supplier: Ambeed
Warning

A477823-10G A477823-1G A477823-5G A477823-25G
77143-762EA 28.83 USD
77143-762 77143-764 77143-766 77686-263
2,6-Dichloro-4-nitroanisole 97%
2,6-Dichloro-4-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₃
MDL Number: MFCD00061129
CAS Number: 17742-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR