N,N'-Bis(p-toluenesulfonyl)hydrazine 98%

Supplier: Ambeed
Danger

A477602-100G A477602-1G A477602-250MG A477602-25G A477602-5G
77143-752EA 378.01 USD
77143-752 77143-754 77143-756 77143-758 77143-760
N,N'-Bis(p-toluenesulfonyl)hydrazine 98%
N,N'-Bis(p-toluenesulfonyl)hydrazine

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₄S₂
MW: 340.42 g/mol
MDL Number: MFCD00025987
CAS Number: 14062-05-6
UN: 1325
ADR: 4.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR