1-Decyl-3-methylimidazolium chloride 96%

Supplier: Ambeed
Warning

Synonyms: 1-decyl-3-methylimidazolium chloride

A476184-25G A476184-5G A476184-100G
77143-744EA 38.11 USD
77143-744 77143-746 77686-260
1-Decyl-3-methylimidazolium chloride 96%
1-Decyl-3-methylimidazolium chloride

Precaución: for Research Use Only

Formula: C₁₄H₂₇ClN₂
MW: 258.83
MDL Number: MFCD03427605
CAS Number: 171058-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR