1-(4-Fluorophenyl)-1H-imidazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A474739-1G A474739-250MG
77143-730EA 159.65 USD
77143-730 77143-732
1-(4-Fluorophenyl)-1H-imidazole-4-carboxylic acid 97%
1-(4-Fluorophenyl)-1H-imidazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇FN₂O₂
MDL Number: MFCD08752952
CAS Number: 114067-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR