4-Fluorophenyl methyl sulfone 97%

Supplier: Ambeed
Warning

Synonyms: 1-Fluoro-4-(methylsulphonyl)benzene, 1-Fluor-4-(methylsulphonyl)benzol

A474498-100G A474498-1G A474498-25G A474498-5G
77143-722EA 229.16 USD
77143-722 77143-724 77143-726 77143-728
4-Fluorophenyl methyl sulfone 97%
4-Fluorophenyl methyl sulfone

Precaución: for Research Use Only

Formula: C₇H₇FO₂S
MW: 174.2 g/mol
MDL Number: MFCD00039753
CAS Number: 455-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR