(4-(Di([1,1'-biphenyl]-4-yl)amino)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A474439-100MG A474439-1G A474439-250MG
77143-716EA 39.26 USD
77143-716 77143-718 77143-720
(4-(Di([1,1'-biphenyl]-4-yl)amino)phenyl)boronic acid 97%
(4-(Di([1,1'-biphenyl]-4-yl)amino)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₃₀H₂₄BNO₂
MW: 441.33 g/mol
MDL Number: MFCD22581306
CAS Number: 943836-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR